Få mere at vide
O,O-Diethyl Thiophosphate Potassium Salt, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | O,O-Diethyl hydrogen thiophosphate potassium salt |
| CAS | 5871-17-0 |
| Molekylær formel | C4 H10 O3 P S . K |
| Mængde | 250 mg |
| Synonym | Phosphorothioic acid, O,O-diethyl ester, potassium salt (1:1), Ethyl potassium phosphorothioate (6CI,7CI), Phosphorothioic acid, O,O-diethyl ester, potassium salt (8CI,9CI), O,O-Diethyl potassium phosphorothioate, Potassium O,O-diethyl phosphorothioate, Potassium O,O-diethylthiophosphate, Potassium di-O-ethyl phosphorothioate, Potassium diethyl thiophosphate |
| InChI formel | InChI=1S/C4H11O3PS.K/c1-3-6-8(5,9)7-4-2;/h3-4H2,1-2H3,(H,5,9);/q;+1/p-1 |
| SMIL | [K+].CCOP(=O)([S-])OCC |
| IUPAC navn | potassium;1-[ethoxy(sulfido)phosphoryl]oxyethane |
| Molekylvægt (g/mol) | 208.26 |
| Formel vægt | 207.9725 |