Få mere at vide
O,O-Diethyl Thiophosphate Ammonium Salt, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | O,O-Diethyl Thiophosphate Ammonium salt |
| CAS | 5871-16-9 |
| Molekylær formel | C4 H10 O3 P S . H4 N |
| Mængde | 100 mg |
| Synonym | Phosphorothioic acid, O,O-diethyl ester, ammonium salt (1:1), Ammonium ethyl phosphorothioate ((NH4O)(EtO)2PS) (7CI), Phosphorothioic acid, O,O-diethyl ester, ammonium salt (8CI,9CI), Ammonium O,O-diethyl phosphorothioate, Ammonium O,O-diethyl thiophosphate |
| InChI formel | InChI=1S/C4H11O3PS.H3N/c1-3-6-8(5,9)7-4-2;/h3-4H2,1-2H3,(H,5,9);1H3 |
| SMIL | [NH4+].CCOP(=O)([S-])OCC |
| IUPAC navn | azanium;1-[ethoxy(sulfido)phosphoryl]oxyethane |
| Molekylvægt (g/mol) | 187.2 |
| Formel vægt | 187.0432 |