missing translation for 'onlineSavingsMsg'
Få mere at vide

O,O-Diethyl Dithiophosphate-13C4 Ammonium Salt, TRC

Artikelnummer. 30379967
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30379967 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30379967 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4442671MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale O,O-Diethyl Dithiophosphate-13C4 Ammonium Salt
CAS 1329641-22-6
Molekylær formel 13C4H14NO2PS2
Mængde 1 mg
Synonym Phosphorodithioic Acid O,O-Diethyl Ester-13C4 Ammonium Salt, Ammonium Ethyl Phosphorodithioate-13C4, Phosphorodithioic Acid O,O-Diethyl Ester-13C4 Ammonium Salt, Ammonium O,O-Diethyl Dithiophosphate-13C4, Ammonium O,O-Diethyl Phosphorodithioate-13C4, Ammonium Di-O-ethyl Dithiophosphate-13C4, Ammonium Diethyldithiophosphate-13C4, NF 133-13C4, O,O-Diethyl Ammonium Phosphorodithioate-13C4, O,O-Diethylphosphorodithioic Acid-13C4 Ammonium Salt
InChI formel InChI=1S/C4H11O2PS2.H3N/c1-3-5-7(8,9)6-4-2;/h3-4H2,1-2H3,(H,8,9);1H3
SMIL SP(O[13CH2][13CH3])(O[13CH2][13CH3])=S.N
Molekylvægt (g/mol) 207.23
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.