missing translation for 'onlineSavingsMsg'
Få mere at vide

O,O'-Bis(trimethylsilyl)-5-fluorouracil, 97%

Artikelnummer. 11359155
Klik for at se tilgængelige muligheder
Mængde:
5 g
25 g
Pakningsstørrelse:
25g
5g
Denne vare kan ikke returneres. Se returpolicy

Artikelnummer. 11359155

Brand: Thermo Scientific Alfa Aesar L16407.14

Request Bulk or Custom Format

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

CAS: 17242-85-2 | C10H19FN2O2Si2 | 274.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 17242-85-2
Molekylær formel C10H19FN2O2Si2
Molekylvægt (g/mol) 274.44
MDL nummer MFCD00042441
InChI nøgle MPVQFVDWJDULDL-UHFFFAOYSA-N
Synonym 2,4-bis trimethylsilyl-5-fluorouracil, o,o'-bis trimethylsilyl-5-fluorouracil, 5-fluoro-2,4-bis trimethylsilyl oxy pyrimidine, 2,4-bis trimethylsilyloxy-5-fluoropyrimidine, 5-fluorouracil, tms, uracil, 5-fluoro, tms, 2,4-bis trimethylsilyl-5-fluorouraci, 5-fluoro-2,4-bis trimethylsilyloxy pyrimidine, 2,4-di-trimethylsilyloxy-5-fluoropyrimidine
PubChem CID 140203
IUPAC navn (5-fluoro-2-trimethylsilyloxypyrimidin-4-yl)oxy-trimethylsilane
SMIL C[Si](C)(C)OC1=NC=C(F)C(O[Si](C)(C)C)=N1

Tekniske data

Kemisk navn eller materiale O,O'-Bis(trimethylsilyl)-5-fluorouracil
Tæthed 1.05
Kogepunkt 114°C (14 mmHg)
Flammepunkt 32°C (89°F)
Brydningsindeks 1.459
Mængde 25 g
FN nummer UN1992
Beilstein 885268
Følsomhed Moisture sensitive
Formel vægt 274.44
Procent renhed 97%
Vis mere Vis mindre

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.