missing translation for 'onlineSavingsMsg'
Learn More

O,O'-Bis(trimethylsilyl)-5-fluorouracil, 97%, Thermo Scientific Chemicals

Catalog Number 11349155
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
17242-85-2
C10H19FN2O2Si2
274.44
MFCD00042441
MPVQFVDWJDULDL-UHFFFAOYSA-N
2,4-bis trimethylsilyl-5-fluorouracil, o,o'-bis trimethylsilyl-5-fluorouracil, 5-fluoro-2,4-bis trimethylsilyl oxy pyrimidine, 2,4-bis trimethylsilyloxy-5-fluoropyrimidine, 5-fluorouracil, tms, uracil, 5-fluoro, tms, 2,4-bis trimethylsilyl-5-fluorouraci, 5-fluoro-2,4-bis trimethylsilyloxy pyrimidine, 2,4-di-trimethylsilyloxy-5-fluoropyrimidine
140203
(5-fluoro-2-trimethylsilyloxypyrimidin-4-yl)oxy-trimethylsilane
C[Si](C)(C)OC1=NC=C(F)C(O[Si](C)(C)C)=N1
This item is not returnable. View return policy
17242-85-2
C10H19FN2O2Si2
274.44
MFCD00042441
MPVQFVDWJDULDL-UHFFFAOYSA-N
2,4-bis trimethylsilyl-5-fluorouracil, o,o'-bis trimethylsilyl-5-fluorouracil, 5-fluoro-2,4-bis trimethylsilyl oxy pyrimidine, 2,4-bis trimethylsilyloxy-5-fluoropyrimidine, 5-fluorouracil, tms, uracil, 5-fluoro, tms, 2,4-bis trimethylsilyl-5-fluorouraci, 5-fluoro-2,4-bis trimethylsilyloxy pyrimidine, 2,4-di-trimethylsilyloxy-5-fluoropyrimidine
140203
(5-fluoro-2-trimethylsilyloxypyrimidin-4-yl)oxy-trimethylsilane
C[Si](C)(C)OC1=NC=C(F)C(O[Si](C)(C)C)=N1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.05
Boiling Point 114°C (14 mmHg)
Flash Point 32°C (89°F)
Refractive Index 1.459
Quantity 5 g
UN Number UN1992
Beilstein 885268
Sensitivity Moisture sensitive
Formula Weight 274.44
Percent Purity 97%
Chemical Name or Material O,O'-Bis(trimethylsilyl)-5-fluorouracil
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'