Få mere at vide
o-Nitrophenyl 2-Acetamido-2-deoxy-Alpha-D-galactopyranoside, TRC
Artikelnummer. 30471217
Skift visning
Mængde:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30471217 10 mg
30470296 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30471217 Leverandør Toronto Research Chemicals Leverandørnr. TRCN49925010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale o-Nitrophenyl 2-Acetamido-2-deoxy-a-D-galactopyranoside
CAS 23646-67-5
Molekylær formel C14 H18 N2 O8
Mængde 10 mg
InChI formel InChI=1S/C14H18N2O8/c1-7(18)15-11-13(20)12(19)10(6-17)24-14(11)23-9-5-3-2-4-8(9)16(21)22/h2-5,10-14,17,19-20H,6H2,1H3,(H,15,18)/t10-,11-,12+,13-,14+/m1/s1
SMIL CC(=O)N[C@@H]1[C@@H](O)[C@@H](O)[C@@H](CO)O[C@@H]1Oc2ccccc2[N+](=O)[O-]
IUPAC navn N-[(2R,3R,4R,5R,6R)-4,5-dihydroxy-6-(hydroxymethyl)-2-(2-nitrophenoxy)oxan-3-yl]acetamide
Molekylvægt (g/mol) 342.3
Formel vægt 342.1063