Få mere at vide
O-Methyl Meloxicam, TRC
Artikelnummer. 30135792
Skift visning
Mængde:
250 mg
2.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30135792 2.5 g
30009585 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30135792 Leverandør Toronto Research Chemicals Leverandørnr. TRCM3211502.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1391051-96-9
Molekylær formel C15 H15 N3 O4 S2
Molekylvægt (g/mol) 365.43
Synonym 2H-1,2-Benzothiazine-3-carboxamide, 4-methoxy-2-methyl-N-(5-methyl-2-thiazolyl)-, 1,1-dioxide, O-Methylmeloxicam, 4-Methoxymeloxicam, 4-Methoxy-2-methyl-N-(5-methyl-1,3-thiazol-2-yl)-2H-1,2-benzothiazine-3-carboxamide 1,1-dioxide
IUPAC navn 4-methoxy-2-methyl-N-(5-methyl-1,3-thiazol-2-yl)-1,1-dioxo-1λ^{6},2-benzothiazine-3-carboxamide
SMIL COC1=C(N(C)S(=O)(=O)c2ccccc12)C(=O)Nc3ncc(C)s3
Tekniske data
Kemisk navn eller materiale 4-Methoxymeloxicam
Mængde 2.5 g
InChI formel InChI=1S/C15H15N3O4S2/c1-9-8-16-15(23-9)17-14(19)12-13(22-3)10-6-4-5-7-11(10)24(20,21)18(12)2/h4-8H,1-3H3,(H,16,17,19)
Formel vægt 365.0504