Få mere at vide
O-Methyl-L-threonine, TRC
Artikelnummer. 30476005
Skift visning
Mængde:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30476005 100 mg
30477232 50 mg
30474897 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30476005 Leverandør Toronto Research Chemicals Leverandørnr. TRCM329983100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 4144-02-9
Molekylær formel C5H11NO3
Molekylvægt (g/mol) 133.15
Synonym (2S,3R)-2-Amino-3-methoxybutanoic Acid, L-O-Methylthreonine, O-Methylthreonine
IUPAC navn (2S,3R)-2-amino-3-methoxybutanoic acid
SMIL CO[C@H](C)[C@H](N)C(=O)O
Tekniske data
Kemisk navn eller materiale O-Methyl-L-threonine
InChI formel InChI=1S/C5H11NO3/c1-3(9-2)4(6)5(7)8/h3-4H,6H2,1-2H3,(H,7,8)/t3-,4+/m1/s1
Mængde 100 mg
Formel vægt 133.0739