Få mere at vide
O-Methyl Atovaquone, TRC
Artikelnummer. 17951495
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17951495 10 mg
17903705 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17951495 Leverandør Toronto Research Chemicals Leverandørnr. TRCM28830010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 129700-41-0
Molekylær formel C23 H21 Cl O3
Molekylvægt (g/mol) 380.86
Synonym 2-[trans-4-(4-Chlorophenyl)cyclohexyl]-3-methoxy-1,4-naphthalenedione, trans-2-[4-(4-Chlorophenyl)cyclohexyl]-3-methoxy-1,4-naphthalenedione, 2-[trans-4-(4-Chlorophenyl)cyclohexyl]-3-methoxynaphthalene-1,4-dione, Atovaquone Imp. D (EP), Trans-2-[4-(4-Chlorophenyl)cyclohexyl]-3-methoxy-1,4-naphthalenedione, 2-((1r,4r)-4-(4-Chlorophenyl)cyclohexyl)-3-methoxynaphthalene-1,4-dione, Atorvastatin EP Impurity D
IUPAC navn 2-[4-(4-chlorophenyl)cyclohexyl]-3-methoxynaphthalene-1,4-dione
SMIL COC1=C([C@@H]2CC[C@H](CC2)c3ccc(Cl)cc3)C(=O)c4ccccc4C1=O
Tekniske data
Kemisk navn eller materiale O-Methyl Atovaquone
Mængde 10 mg
InChI formel InChI=1S/C23H21ClO3/c1-27-23-20(21(25)18-4-2-3-5-19(18)22(23)26)16-8-6-14(7-9-16)15-10-12-17(24)13-11-15/h2-5,10-14,16H,6-9H2,1H3/t14-,16-
Formel vægt 380.1179