missing translation for 'onlineSavingsMsg'
Få mere at vide

o-hydroxy-alfa, alfa-dimethylbenzylalkohol, TRC

Artikelnummer. 30408294
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30408294 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30408294 Leverandør Toronto Research Chemicals Leverandørnr. TRCH9413801G
 Begrænset vare
Forventet forsendelse: 10-12-2026
Læg i indkøbskurv
Læg i indkøbskurv

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

Højrenheds organiske molekyler og analytiske standarder, strategisk leveret verden over for at muliggøre innovation og kommerciel succes.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale o-Hydroxy-alpha,alpha-dimethylbenzyl Alcohol
CAS 3045-32-7
Molekylær formel C9H12O2
Mængde 1 g
Synonym 2-Hydroxy-alpha,alpha-dimethylbenzenemethanol
InChI formel InChI=1S/C9H12O2/c1-9(2,11)7-5-3-4-6-8(7)10/h3-6,10-11H,1-2H3
SMIL CC(C)(O)c1ccccc1O
IUPAC navn 2-(2-hydroxypropan-2-yl)phenol
Molekylvægt (g/mol) 152.19
Formel vægt 152.0837
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.