Få mere at vide
O-Desmethylsulpiride, LoGiCal
Artikelnummer. 30463017
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30463017 10 mg
1 options
Artikelnummer. 30463017 Leverandør LoGiCal Leverandørnr. LGCFOR0051.07
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale O-Desmethylsulpiride
CAS 67381-52-6
Analyt- eller komponentnavne O-Desmethylsulpiride
Forsendelsestilstand Room Temperature
Molekylær formel C14 H21 N3 O4 S
Certificeringer/compliance ISO 9001
Synonym (RS)-N-[(1-Ethylpyrrolidin-2-yl)methyl]-2-hydroxy-5-sulphamoylbenzamide, Sulpiride Imp. G (EP), O-Desmethylsulpiride, Sulpiride Impurity G
InChI formel InChI=1S/C14H21N3O4S/c1-2-17-7-3-4-10(17)9-16-14(19)12-8-11(22(15,20)21)5-6-13(12)18/h5-6,8,10,18H,2-4,7,9H2,1H3,(H,16,19)(H2,15,20,21)
Løsningstype Neat
SMIL CCN1CCCC1CNC(=O)c2cc(ccc2O)S(=O)(=O)N
IUPAC navn N-[(1-ethylpyrrolidin-2-yl)methyl]-2-hydroxy-5-sulfamoylbenzamide
Industry Type Clinical Testing
Molekylvægt (g/mol) 327.4
Mængde 10 mg
Formel vægt 327.1253 g/mol