missing translation for 'onlineSavingsMsg'
Få mere at vide

(-)-O-Desmethyl-N,N-bisdesmethyl Tramadol, TRC

Artikelnummer. 30353073
Change view
Click to view available options
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30353073 1 mg
30351689 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30353073 Supplier Toronto Research Chemicals Supplier No. TRCD2911501MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 185502-41-4
Molekylær formel C13H19NO2
Molekylvægt (g/mol) 221.3
Synonym 3-[(1S,2S)-2-(Aminomethyl)-1-hydroxycyclohexyl]phenol
SMIL OC1=CC=CC([C@]2(O)CCCC[C@@]2([H])CN)=C1

Specifications

Kemisk navn eller materiale (-)-O-Desmethyl-N,N-bisdesmethyl Tramadol
Mængde 1 mg
InChI formel InChI=1S/C13H19NO2/c14-9-11-4-1-2-7-13(11,16)10-5-3-6-12(15)8-10/h3,5-6,8,11,15-16H,1-2,4,7,9,14H2/t11-,13+/m0/s1
Formel vægt 221.1416
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.