Få mere at vide
O-Desmethyl Mycophenolic Acid, TRC
Artikelnummer. 30360989
Skift visning
Mængde:
1 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30360989 1 mg
30358560 10 mg
30362329 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30360989 Leverandør Toronto Research Chemicals Leverandørnr. TRCD2920781MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 31858-65-8
Molekylær formel C16 H18 O6
Molekylvægt (g/mol) 306.31
IUPAC navn (E)-6-(4,6-dihydroxy-7-methyl-3-oxo-1H-2-benzofuran-5-yl)-4-methylhex-4-enoic acid
SMIL C\C(=C/Cc1c(O)c(C)c2COC(=O)c2c1O)\CCC(=O)O
Tekniske data
Kemisk navn eller materiale O-Desmethyl Mycophenolic Acid
Mængde 1 mg
InChI formel InChI=1S/C16H18O6/c1-8(4-6-12(17)18)3-5-10-14(19)9(2)11-7-22-16(21)13(11)15(10)20/h3,19-20H,4-7H2,1-2H3,(H,17,18)/b8-3+
Formel vægt 306.1103