Få mere at vide
O-Desmethyl Metoprolol-d5, TRC
Artikelnummer. 30355899
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30355899 10 mg
30350916 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30355899 Leverandør Toronto Research Chemicals Leverandørnr. TRCD29207210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1189981-81-4
Molekylær formel C14H18D5NO3
Molekylvægt (g/mol) 258.37
Synonym 4-[2-Hydroxy-3-[(1-methylethyl)amino]propoxy]-benzeneethanol-d5, (+/-)-4-[2-Hydroxy-3-(isopropylamino)propoxy]phenylethyl Alcohol-d5, (+/-)-O-Demethylmetoprolol-d5, H 105/22-d5, SL 80-0088-d5
SMIL [2H]C([2H])(NC(C)C)C([2H])(O)C([2H])([2H])Oc1ccc(CCO)cc1
Tekniske data
Kemisk navn eller materiale O-Desmethyl Metoprolol-d5
Mængde 10 mg
InChI formel InChI=1S/C14H23NO3/c1-11(2)15-9-13(17)10-18-14-5-3-12(4-6-14)7-8-16/h3-6,11,13,15-17H,7-10H2,1-2H3/i9D2,10D2,13D