Få mere at vide
O-Desmethyl Indomethacin-d4, TRC
Artikelnummer. 30361008
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30361008 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30361008 Leverandør Toronto Research Chemicals Leverandørnr. TRCD2918021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale O-Desmethyl Indomethacin-d4
CAS 1189916-55-9
Molekylær formel C18H10D4ClNO4
Mængde 1 mg
Synonym 1-(4-Chlorobenzoyl-d4)-5-hydroxy-2-methyl-1H-indole-3-acetic Acid, 5-Hydroxy-
InChI formel InChI=1S/C18H14ClNO4/c1-10-14(9-17(22)23)15-8-13(21)6-7-16(15)20(10)18(24)11-2-4-12(19)5-3-11/h2-8,21H,9H2,1H3,(H,22,23)/i2D,3D,4D,5D
SMIL O=C(C1=C([2H])C([2H])=C(C([2H])=C1[2H])Cl)N2C3=CC=C(O)C=C3C(CC(O)=O)=C2C
IUPAC navn 2-(1-(4-chlorobenzoyl-2,3,5,6-d4)-5-hydroxy-2-methyl-1H-indol-3-yl)acetic acid
Molekylvægt (g/mol) 347.79
Formel vægt 347.09