Få mere at vide
O-Desmethyl Carvedilol, TRC
Artikelnummer. 30137932
Skift visning
Mængde:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30137932 25 mg
30007113 10 mg
30012674 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30137932 Leverandør Toronto Research Chemicals Leverandørnr. TRCD29147525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 72956-44-6
Molekylær formel C23H24N2O4
Molekylvægt (g/mol) 392.45
Synonym Phenol, 2-[2-[[3-(9H-carbazol-4-yloxy)-2-hydroxypropyl]amino]ethoxy]-, 2-[2-[[3-(9H-Carbazol-4-yloxy)-2-hydroxypropyl]amino]ethoxy]phenol, BM 14242, Desmethylcarvedilol, O-Desmethylcarvedilol
SMIL OC(CNCCOc1ccccc1O)COc2cccc3[nH]c4ccccc4c23
Tekniske data
Kemisk navn eller materiale O-Desmethyl Carvedilol
Mængde 25 mg
InChI formel InChI=1S/C23H24N2O4/c26-16(14-24-12-13-28-21-10-4-3-9-20(21)27)15-29-22-11-5-8-19-23(22)17-6-1-2-7-18(17)25-19/h1-11,16,24-27H,12-15H2
Formel vægt 392.1736