Få mere at vide
O-Desmethyl Carvedilol-d5, TRC
Artikelnummer. 30353531
Skift visning
Mængde:
2.5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30353531 2.5 mg
30352118 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30353531 Leverandør Toronto Research Chemicals Leverandørnr. TRCD2914772.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1794817-24-5
Molekylær formel C23H19D5N2O4
Molekylvægt (g/mol) 397.48
Synonym 2-[2-[[3-(9H-Carbazol-4-yloxy)-2-hydroxy(propyl-d5)]amino]ethoxy]phenol
IUPAC navn 2-(2-((3-((9H-carbazol-4-yl)oxy)-2-hydroxypropyl-1,1,2,3,3-d5)amino)ethoxy)phenol
SMIL O(C(C(C(NCCOC1=C(O)C=CC=C1)([2H])[2H])(O)[2H])([2H])[2H])C2=C3C=4C(NC3=CC=C2)=CC=CC4
Tekniske data
Kemisk navn eller materiale O-Desmethyl Carvedilol-d5
Mængde 2.5 mg
InChI formel InChI=1S/C23H24N2O4/c26-16(14-24-12-13-28-21-10-4-3-9-20(21)27)15-29-22-11-5-8-19-23(22)17-6-1-2-7-18(17)25-19/h1-11,16,24-27H,12-15H2/i14D2,15D2,16D
Formel vægt 397.2