Få mere at vide
O-Desisopropyl-O-methoxyethyl Nimodipine, TRC
Artikelnummer. 30006796
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30006796 100 mg
30005484 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30006796 Leverandør Toronto Research Chemicals Leverandørnr. TRCD290165100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 70172-96-2
Molekylær formel C21 H26 N2 O8
Molekylvægt (g/mol) 434.44
Synonym Bis(2-methoxyethyl) 2,6-Dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate, Nimodipine Imp. C (EP)
IUPAC navn bis(2-methoxyethyl) 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate
SMIL COCCOC(=O)C1=C(C)NC(=C(C1c2cccc(c2)[N+](=O)[O-])C(=O)OCCOC)C
Tekniske data
Kemisk navn eller materiale Bis(2-methoxyethyl) 2,6-Dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate
Renhedsgrad noter HPLC
Mængde 100 mg
InChI formel InChI=1S/C21H26N2O8/c1-13-17(20(24)30-10-8-28-3)19(15-6-5-7-16(12-15)23(26)27)18(14(2)22-13)21(25)31-11-9-29-4/h5-7,12,19,22H,8-11H2,1-4H3
Formel vægt 434.1689
Procent renhed >95%