Få mere at vide
O-Desethyl-O-propyl Methisosildenafil, TRC
Artikelnummer. 30365880
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30365880 100 mg
30355061 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30365880 Leverandør Toronto Research Chemicals Leverandørnr. TRCD289495100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1391053-82-9
Molekylær formel C24 H34 N6 O4 S
Molekylvægt (g/mol) 502.6296
Synonym rel-5-[5-[[(3R,5S)-3,5-Dimethyl-1-piperazinyl]sulfonyl]-2-propoxyphenyl]-1,6-dihydro-1-methyl-3-propyl-7H-pyrazolo[4,3-d]pyrimidin-7-one, Propoxyphenyl Aildenafil
IUPAC navn 5-[5-[(3R,5S)-3,5-dimethylpiperazin-1-yl]sulfonyl-2-propoxyphenyl]-1-methyl-3-propyl-4H-pyrazolo[4,3-d]pyrimidin-7-one
SMIL CCCOc1ccc(cc1C2=NC(=O)c3c(N2)c(CCC)nn3C)S(=O)(=O)N4C[C@@H](C)N[C@@H](C)C4
Tekniske data
Kemisk navn eller materiale Propoxyphenyl Aildenafil
Mængde 100 mg
InChI formel InChI=1S/C24H34N6O4S/c1-6-8-19-21-22(29(5)28-19)24(31)27-23(26-21)18-12-17(9-10-20(18)34-11-7-2)35(32,33)30-13-15(3)25-16(4)14-30/h9-10,12,15-16,25H,6-8,11,13-14H2,1-5H3,(H,26,27,31)/t15-,16+
Formel vægt 502.2362