Få mere at vide
o-Cresol-d3 (methyl-d3), TRC
Artikelnummer. 30427982
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30427982 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30427982 Leverandør Toronto Research Chemicals Leverandørnr. TRCO78191250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Methylphenol-D3 (methyl-D3)
CAS 70837-27-3
Molekylær formel C7H5D3O
Mængde 50 mg
Synonym Phenol','2-(methyl-d3)- (9CI','ACI), 2-(Methyl-d3)phenol (ACI), o-Cresol-d3 (methyl-d3), 2-Methylphenol-D3 (methyl-D3)
InChI formel InChI=1S/C7H8O/c1-6-4-2-3-5-7(6)8/h2-5,8H,1H3/i1D3
SMIL OC1=CC=CC=C1C([2H])([2H])[2H]
IUPAC navn 2-(methyl-d3)phenol
Molekylvægt (g/mol) 111.16
Formel vægt 111.08