missing translation for 'onlineSavingsMsg'
Learn More

o-Cresol, ≥99, Reagent Grade, Honeywell™

Brand:  Honeywell Research Chemicals C85700-5G

Additional Details : CAS Number : 95-48-7 Weight : 0.00500kg

Packaging Glass bottle
Quantity 5g

 View more versions of this product

Product Code. 15637780

  • 148.79 DKK / 5g


Estimated Shipment date
from Supplier 25-07-2018
Add to basket

Specifications

Chemical Identifiers

3.72 (vs air)
1110°F
MFCD00002226
≥99%
95-48-7
Solid
Characteristic
191°C at 1,013 hPa
26g/L at 25°C
81°C (177.8°F)
CH3C6H4OH
UN3455
o-Cresol
506917
Colorless
30.99°C at 1,013 hPa
1.05g/cu.cm at 20°C

Specifications

o-Cresol
108.14g/mol
95-48-7
Characteristic
≥99%
CH3C6H4OH
MFCD00002226
506917
Solid
CC1=CC=CC=C1O
108.14
191°C at 1,013 hPa
Colorless
1110°F
81°C (177.8°F)
5g
1.05g/cu.cm at 20°C
Glass bottle
C7H8O
26g/L at 25°C
UN3455
3.72 (vs air)
QWVGKYWNOKOFNN-UHFFFAOYSA-N
2-methylphenol
335
CHEBI:28054
30.99°C at 1,013 hPa
Videos