Få mere at vide
o-Carbomethoxybenzyl Sulfonamide, TRC
Artikelnummer. 30364638
Skift visning
Mængde:
5 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30364638 25 g
30354372 10 g
30355667 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30364638 Leverandør Toronto Research Chemicals Leverandørnr. TRCC17772825G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 112941-26-1
Molekylær formel C9H11NO4S
Molekylvægt (g/mol) 229.25
Synonym 2-(Methoxycarbonyl)benzylsulfonamide, Methyl 2-(sulfamoylmethyl)benzoate, Methyl 2-[(aminosulfonyl)methyl]benzoate, [2-(Methoxycarbonyl)phenyl]methanesulfonamide
IUPAC navn methyl 2-(sulfamoylmethyl)benzoate
SMIL COC(=O)c1ccccc1CS(=O)(=O)N
Tekniske data
Kemisk navn eller materiale o-Carbomethoxybenzyl Sulfonamide
Mængde 25 g
InChI formel InChI=1S/C9H11NO4S/c1-14-9(11)8-5-3-2-4-7(8)6-15(10,12)13/h2-5H,6H2,1H3,(H2,10,12,13)
Formel vægt 229.0409