Få mere at vide
O-Benzyl-S-(methyl-d3)-3-thioacetaminophen, TRC
Artikelnummer. 30337366
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30337366 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30337366 Leverandør Toronto Research Chemicals Leverandørnr. TRCB285752100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale O-Benzyl-S-(methyl-d3)-3-thioacetaminophen
CAS 1184990-56-4
Molekylær formel C16H14D3NO2S
Mængde 100 mg
Synonym 4-Benzoxy-3-(methylthio-d3)acetanilide
InChI formel InChI=1S/C16H17NO2S/c1-12(18)17-14-8-9-15(16(10-14)20-2)19-11-13-6-4-3-5-7-13/h3-10H,11H2,1-2H3,(H,17,18)/i2D3
SMIL [2H]C([2H])([2H])Sc1cc(NC(=O)C)ccc1OCc2ccccc2
IUPAC navn N-[4-phenylmethoxy-3-(trideuteriomethylsulfanyl)phenyl]acetamide
Molekylvægt (g/mol) 290.4
Formel vægt 290.1168