Få mere at vide
O-Arachidonoyl Ethanolamine Hydrochloride (Technical Grade), TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | O-Arachidonoyl Ethanolamine (hydrochloride) |
| CAS | 443129-35-9 |
| Molekylær formel | C22 H37 N O2 . H Cl |
| Mængde | 1 mg |
| Synonym | 5,8,11,14-Eicosatetraenoic acid, 2-aminoethyl ester, hydrochloride (1:1), (5Z,8Z,11Z,14Z)- (ACI), 5,8,11,14-Eicosatetraenoic acid, 2-aminoethyl ester, hydrochloride, (5Z,8Z,11Z,14Z)- (9CI), O-Arachidonoyl ethanolamine hydrochloride, Virodhamine hydrochloride, (5Z,8Z,11Z,14Z)-5,8,11,14-Eicosatetraenoic acid 2-aminoethyl ester hydrochloride |
| InChI formel | InChI=1S/C22H37NO2.ClH/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-22(24)25-21-20-23;/h6-7,9-10,12-13,15-16H,2-5,8,11,14,17-21,23H2,1H3;1H/b7-6-,10-9-,13-12-,16-15-; |
| SMIL | Cl.CCCCC\C=C/C\C=C/C\C=C/C\C=C/CCCC(=O)OCCN |
| IUPAC navn | 2-aminoethyl (5Z,8Z,11Z,14Z)-icosa-5,8,11,14-tetraenoate;hydrochloride |
| Molekylvægt (g/mol) | 384 |
| Formel vægt | 383.2591 |