Learn More
o-Anisidine, TRC
Product Code. 30137328
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137328 5 g 5g
18053010 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.
Product Code. 30137328 Supplier Toronto Research Chemicals Supplier No. TRCA6735205G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 90-04-0
Molecular Formula C7 H9 N O
Molecular Weight (g/mol) 123.15
Synonym o-Anisidine (8CI), 2-Methoxybenzenamine, 1-Amino-2-methoxybenzene, 2-Aminoanisole, 2-Aminomethoxybenzene, 2-Methoxy-1-aminobenzene, 2-Methoxyaniline, 2-Methoxyphenylamine, NSC 3122, o-Aminoanisole, o-Aminomethoxybenzene, o-Methoxyaniline, o-Methoxyphenylamine
IUPAC Name 2-methoxyaniline
SMILES COc1ccccc1N
Specifications
Quantity 5 g
InChI Formula InChI=1S/C7H9NO/c1-9-7-5-3-2-4-6(7)8/h2-5H,8H2,1H3
Formula Weight 123.0684
Chemical Name or Material 2-Anisidine (2-Methoxyaniline)