Få mere at vide
o-Anisidine-d3, TRC
Artikelnummer. 30336942
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30336942 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30336942 Leverandør Toronto Research Chemicals Leverandørnr. TRCA673523100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale o-Anisidine methyl-d3
CAS 1398066-00-6
Molekylær formel C7 D3 H6 N O
Mængde 100 mg
Synonym 2-Methoxy-d3-aniline, 1-Amino-2-(methoxy-d3)benzene, o-Anisidine-d3, 2-Methoxyaniline-D3 (methoxy-D3), 2-Methoxyaniline D3 (methoxy D3), 2-Anisidine D3 (methoxy D3)
InChI formel InChI=1S/C7H9NO/c1-9-7-5-3-2-4-6(7)8/h2-5H,8H2,1H3/i1D3
SMIL [2H]C([2H])([2H])Oc1ccccc1N
IUPAC navn 2-(trideuteriomethoxy)aniline
Molekylvægt (g/mol) 126.17
Formel vægt 126.087