Få mere at vide
O-Acetylmetronidazole, Mikromol™
Artikelnummer. 17930546
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
17930546 25 mg
1 options
Artikelnummer. 17930546 Leverandør Mikromol Leverandørnr. MM0012.110025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
O-Acetylmetronidazole
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Metronidazole Acetate
CAS 13182-82-6
Analyt- eller komponentnavne Metronidazole Acetate
Forsendelsestilstand Room Temperature
Molekylær formel C8H11N3O4
Synonym O-Acetylmetronidazole, 2-Methyl-5-nitro-1H-imidazole-1-ethanol 1-Acetate, 2-Methyl-5-nitro-imidazole-1-ethanol Acetate
SMIL CC(=O)OCCn1c(C)ncc1[N+](=O)[O-]
Molekylvægt (g/mol) 213.19