Få mere at vide
o-Acetyllamivudine, TRC
Artikelnummer. 30317561
Skift visning
Mængde:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30317561 50 mg
30326210 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30317561 Leverandør Toronto Research Chemicals Leverandørnr. TRCA26981050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale O-Acetyllamivudine
CAS 158704-07-5
Molekylær formel C10 H13 N3 O4 S
Mængde 50 mg
Synonym o-Acetyllamivudine, 2(1H)-Pyrimidinone, 1-[(2R,5S)-2-[(acetyloxy)methyl]-1,3-oxathiolan-5-yl]-4-amino-, 1-[(2R,5S)-2-[(Acetyloxy)methyl]-1,3-oxathiolan-5-yl]-4-amino-2(1H)-pyrimidinone, 2(1H)-Pyrimidinone, 1-[2-[(acetyloxy)methyl]-1,3-oxathiolan-5-yl]-4-amino-, (2R-cis)-, O-Acetyllamivudine
InChI formel InChI=1S/C10H13N3O4S/c1-6(14)16-4-9-17-8(5-18-9)13-3-2-7(11)12-10(13)15/h2-3,8-9H,4-5H2,1H3,(H2,11,12,15)/t8-,9+/m0/s1
SMIL CC(=O)OC[C@@H]1O[C@@H](CS1)N2C=CC(=NC2=O)N
IUPAC navn [(2R,5S)-5-(4-amino-2-oxopyrimidin-1-yl)-1,3-oxathiolan-2-yl]methyl acetate
Molekylvægt (g/mol) 271.293
Formel vægt 271.063