Få mere at vide
O-Acetyl-L-serine, TRC
Artikelnummer. 30414028
Skift visning
Mængde:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30414028 100 mg
30410816 500 mg
30411926 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30414028 Leverandør Toronto Research Chemicals Leverandørnr. TRCO298210100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 5147-00-2
Molekylær formel C5H9NO4
Molekylvægt (g/mol) 147.13
Synonym (S)-3-Acetoxy-2-aminopropanoic Acid, NSC 226230, O-Acetylserine
IUPAC navn (2S)-3-acetyloxy-2-aminopropanoic acid
SMIL CC(=O)OC[C@H](N)C(=O)O
Tekniske data
Kemisk navn eller materiale O-Acetyl-L-serine
InChI formel InChI=1S/C5H9NO4/c1-3(7)10-2-4(6)5(8)9/h4H,2,6H2,1H3,(H,8,9)/t4-/m0/s1
Mængde 100 mg
Formel vægt 147.0532