Få mere at vide
O-6-Methyl-2'-deoxyguanosine-D3, TRC
Artikelnummer. 30467153
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30467153 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30467153 Leverandør Toronto Research Chemicals Leverandørnr. TRCM3847921MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale O-6-Methyl-2'-deoxyguanosine-D3
CAS 99104-20-8
Molekylær formel C11 H12 D3 N5 O4
Mængde 1 mg
InChI formel InChI=1S/C11H15N5O4/c1-19-10-8-9(14-11(12)15-10)16(4-13-8)7-2-5(18)6(3-17)20-7/h4-7,17-18H,2-3H2,1H3,(H2,12,14,15)/t5-,6+,7+/m0/s1/i1D3
SMIL [2H]C([2H])([2H])Oc1nc(N)nc2c1ncn2[C@H]3C[C@H](O)[C@@H](CO)O3
IUPAC navn (2R,3S,5R)-5-[2-amino-6-(trideuteriomethoxy)purin-9-yl]-2-(hydroxymethyl)oxolan-3-ol
Molekylvægt (g/mol) 284.29
Formel vægt 284.1312