Få mere at vide
O-1918, Tocris Bioscience™
Beskrivelse
Selective, silent antagonist of a putative endothelial anandamide receptor distinct from CB1 or CB2 receptors. Inhibits vasodilation and cell migration induced by abnormal-cannabidiol (abn-CBD; Cat. No. 1297).
Tekniske data
Tekniske data
| Kemisk navn eller materiale | O-1918 |
| CAS | 536697-79-7 |
| Molekylær formel | C19H26O2 |
| Mængde | 10 mg |
| Synonym | 1,3-dimethoxy-5-methyl-2-1r,6r-3-methyl-6-1-methylethenyl-2-cyclohexen-1-yl benzene, 1,3-dimethoxy-5-methyl-2-1r,6r-3-methyl-6-1-methylethenyl-2-cyclohexen-1-yl-benzene, 1,3-dimethoxy-5-methyl-2-1r,6r-3-methyl-6-prop-1-en-2-yl cyclohex-2-en-1-yl benzene, benzene,1,3-dimethoxy-5-methyl-2-1r,6r-3-methyl-6-1-methylethenyl-2-cyclohexen-1-yl |
| InChI nøgle | ICHJMVMWPKLUKT-JKSUJKDBSA-N |
| SMIL | CC1=CC(C(CC1)C(=C)C)C2=C(C=C(C=C2OC)C)OC |
| IUPAC navn | 1,3-dimethoxy-5-methyl-2-[(1R,6R)-3-methyl-6-prop-1-en-2-ylcyclohex-2-en-1-yl]benzene |
| Molekylvægt (g/mol) | 286.415 |
| PubChem CID | 40469923 |
| Vis mere |