missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NXNL2 Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP3-06016R
Additional Details : Weight : 0.00970kg
Description
NXNL2 Polyclonal specifically detects NXNL2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
NXNL2 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
C9orf121, chromosome 9 open reading frame 121, nucleoredoxin-like 2, nucleoredoxin-like protein 2, RDCVF2 | |
Produced in rabbits immunized with E. coli-derived Human NXNL2 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 550 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
158046 | |
Store at 4°C in the dark. | |
IgG |