Få mere at vide
NVP-BHG712, MedChemExpress
Artikelnummer. 16435598
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16435598 10 mg
16421767 50 mg
16408297 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 16435598 Leverandør Medchem Express Leverandørnr. HY13258A
MedChemExpress NVP-BHG712 is an oral active EphB4 kinase autophosphorylation inhibitor, with IC50 values of 3.3 nM and 3.0 nM for EphA2 and EphB4, respectively.
NVP-BHG712 is an oral active EphB4 kinase autophosphorylation inhibitor, with IC50 values of 3.3 nM and 3.0 nM for EphA2 and EphB4, respectively.
Tekniske data
Kemisk navn eller materiale NVP-BHG712
Fysisk form Solid
CAS 940310-85-0
Farve White
Mængde 10 mg
Renhedsgrad noter Research
Molekylær formel C26H20F3N7O
Til brug med (applikation) Cancer-Kinase/protease
Synonym BHG712
Opløselighedsinformation DMSO : 31.25 mg/mL (62.07 mM; Need ultrasonic)
SMIL CC1=CC=C(C=C1NC2=NC(C3=CC=CN=C3)=NC4=C2C=NN4C)C(NC5=CC=CC(C(F)(F)F)=C5)=O
Molekylvægt (g/mol) 503.48
Formel vægt 503.48
Procent renhed 99.7%