missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ NUTM1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA559171
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Immunogen sequence: TCPLNVHSYD PQGEGRVDPD LSKPKNLAPL QESQESYTTG TPKATSSHQG LGSTLPRRGT RNAIVPRETS VSKTHRSA Highest antigen sequence identity to the following orthologs: Mouse - 51%, Rat - 53%.
Plays a role in the regulation of proliferation. Regulates TERT expression by modulating SP1 binding to TERT promoter binding sites.
Tekniske data
| NUTM1 | |
| Polyclonal | |
| Unconjugated | |
| NUTM1 | |
| C15orf55; FAM22H; Nuclear protein in testis; NUT; NUT family member 1; NUT midline carcinoma family member 1; NUT midline carcinoma, family member 1; NUTM1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 256646 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.20 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q86Y26 | |
| NUTM1 | |
| Recombinant protein corresponding to Human NUTM1. Recombinant protein control fragment (Product #RP-98181). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion