Få mere at vide
Nutlin 3a, Tocris Bioscience™
MDM2 antagonist; active enantiomer of Nutlin-3 (Cat. No. 3984)
Brand: Tocris Bioscience 6075/25
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Nutlin 3a is a MDM2 antagonist (IC50 = 90 nM); inhibits the MDM2-p53 interaction. Active enantiomer of Nutlin-3 (Cat. No. 3984). Induces expression for P53 regulated genes. Suppresses tumor growth in vivo >98% in prostate cancer and osteosarcoma cancer models. Antiproliferative.Tekniske data
| 4-[[(4S,5R)-4,5-Bis(4-chlorophenyl)-4,5-dihydro-2-[4-methoxy-2-(1-methylethoxy)phenyl]-1H-imidazol-1-yl]carbonyl]-2-piperazinone | |
| 675576-98-4 | |
| >97% | |
| 25 mg |