missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies (±)-Nutlin-3
Description
- (±)-Nutlin-3 is a small-imidazoline-based mouse double minute 2 (MDM2) protein antagonist which disrupts MDM2-p53 interaction (IC50 = 0.09 μM; Vassilev et al.) MDM2 binds the p53 tumor suppressor protein with high affinity and negatively modulates its transcriptional activity and stability
- By disrupting this interaction, (±)-Nutlin-3 therefore promotes the expression of p53-regulated genes and exhibits potent antiproliferative activity in cells with functional p53, but not in cells with mutated p53 (Vassilev et al.), Gu et al.)
Specifications
Specifications
| CAS | 548472-68-0 |
| Mængde | 10 mg |
| Molekylær formel | C30H30Cl2N4O4 |
| Renhed | >98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?