Få mere at vide
Thermo Scientific™ Nunc™ Deep Form Bacteriology Petri Dishes
Artikelnummer. 15367036
Diameter (metrisk):
60 mm
100 mm
150 mm
Pakningsstørrelse:
Pakke med 12
Pakke med 300
Pakke med 400
Pakke med 72
Artikelnummer. 15367036
Brand: Thermo Scientific™ 4031
Culture fungi, bacteria and other microorganisms with these petri dishes
Culture fungi, bacteria and other microorganisms with Thermo Scientific™ Nunc Lab-Tek Disposable Petri Dishes.
Thermo Scientific Nunc Cell Culture products offer solutions for cell growth and analysis and have been used by researchers worldwide since 1953. With a wide range of surface areas and treatments, we have products for all your basic or advanced cell-based research.
  • Promote larger plaques used for efficient library screening
  • Compatible with automated systems
  • Sterile; disposable
  • Deep Petri dishes allow for longer culture periods
Ordering Information:
12 per sealed plastic sleeve.
TRUSTED_SUSTAINABILITY
Tekniske data
Lukketype Stackable
Bund form Flat
Engangs Single-use
Materiale Polystyrene
Form Round
Sterilitet Sterile
Type Petri Dish
Nå tælle 0
nr. pr. sag 300
nr. pr. pakke 12
Aftræk Yes
Højde (metrisk) 26 mm
Højde (engelsk) 26 in.
Diameter (metrisk) 100 mm