missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Thermo Scientific™ Nunc™ 384-Well Clear Polystyrene Plates with Cell Culture Treated and Nontreated Surfaces, 384W Clear Polystyrene, Cell Culture Treated, without lid, Sterile
Beskrivelse
Reduce the volume of reagent needed with Thermo Scientific™ Nunc™ 384-Well Clear Polystyrene Plates with Cell Culture Treated and Nontreated Surfaces, which are optimized for robotics, plate readers and liquid handling systems. 384-well microplates allow sample volume reduction, providing cost savings and allowing more samples to be screened.
Features:
- Clear Polystyrene has excellent optical properties, making it ideal for optical measurements
- Untreated Polystyrene is hydrophobic and suited for growth of suspension cultures that can proliferate and grow without attachment
- Untreated surfaces ideal for adding coatings in the lab
- Cell culture treated, hydrophilic surface promotes cell attachment and growth
- Customized bar coding is available
Tekniske data
Tekniske data
| Certificeringer/compliance | All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) |
| Format | Standard |
| Låg | Without Lid |
| Kulturområde | 0.084 cm2/well |
| Oversigt | Nunclon Delta Treated; no lid; Sterile; 25/100 |
| Array | 16 x 24 |
| Emballage | 25 Pk., 100 Cases |
| Produktlinje | Nunc |
| Farve | Klar |
| Produkttype | Tissue Culture Plate |
| Vis mere |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?