Få mere at vide
Novus Biologicals™ NuMA Recombinant Protein Antigen
Artikelnummer. 18245355 Shop alle Bio Techne produkter
Skift visning
Protein længde:
IRFLELQKVASSSSGNNFLSGSPASPMGDILQTPQFQMRRLKKQLADERSNRDELELELAENRKLLTEKDAQIAMMQQRIDRLALLNEKQAASPLEPKEL
TEEGQQILKQPVSERLDFVCSFLQKNRKHPSSPECLVSAQKVLEGSELELAKMTMLLLYHSTMSSKSPRDWEQFEYKIQAELAVILKFVLDHEDGLNLNEDLENFLQKAPVPSTCSST
Pakningsstørrelse:
0.1ml
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Protein længde unitSize
18245355 TEEGQQILKQPVSERLDFVCSFLQKNRKHPSSPECLVSAQKVLEGSELELAKMTMLLLYHSTMSSKSPRDWEQFEYKIQAELAVILKFVLDHEDGLNLNEDLENFLQKAPVPSTCSST 0.1ml
18235345 IRFLELQKVASSSSGNNFLSGSPASPMGDILQTPQFQMRRLKKQLADERSNRDELELELAENRKLLTEKDAQIAMMQQRIDRLALLNEKQAASPLEPKEL 0.1ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 18245355 Leverandør Novus Biologicals™ Leverandørnr. NBP254673PEP
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human NuMA The NuMA Recombinant Protein Antigen is derived from E. coli. The NuMA Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.
TRUSTED_SUSTAINABILITY
Tekniske data
Protein længde TEEGQQILKQPVSERLDFVCSFLQKNRKHPSSPECLVSAQKVLEGSELELAKMTMLLLYHSTMSSKSPRDWEQFEYKIQAELAVILKFVLDHEDGLNLNEDLENFLQKAPVPSTCSST
Oprensningsmetode >80% by SDS-PAGE and Coomassie blue staining
Fælles navn NUMA1 Recombinant Protein Antigen
Indhold og opbevaring Store at −20°C long term. Avoid freeze-thaw cycles.
Formulering PBS and 1M Urea, pH 7.4
Til brug med (applikation) AC
Gene Alias centrophilin stabilizes mitotic spindle in mitotic cells, nuclear mitotic, apparatus protein 1, NuMA protein
Gen symbol NUMA1
Etikettype Unlabeled
Molekylvægt (g/mol) 29 kDa
Produkttype Recombinant Protein Antigen
Mængde 0.1 mL
Regulatorisk status RUO
Kilde E.coli
Specifik reaktivitet Human
Vis mere Vis mindre
For Research Use Only.