missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nuf2 Rabbit anti-Human, Clone: 5J0T9, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Bio-Techne NBP3-15540-20UL
Additional Details : Weight : 0.00970kg
Description
Nuf2 Monoclonal antibody specifically detects Nuf2 in Human samples. It is validated for Western BlotSpecifications
Nuf2 | |
Monoclonal | |
Unconjugated | |
cancer/testis antigen 106, CDCA1hsNuf2, cell division cycle associated 1, Cell division cycle-associated protein 1, CT106, hNuf2, kinetochore protein Nuf2, NUF2, NDC80 kinetochore complex component, homolog (S. cerevisiae), NUF2RhNuf2R | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Nuf2 (Q9BZD4). | |
20 μg | |
Cell Cycle and Replication | |
83540 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
5J0T9 | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |