Få mere at vide
Macherey-Nagel™ NucleoVac Valves
Mængde:
24 Each
Pakningsstørrelse:
Pakke med 24
Beskrivelse
The NucleoVac 24 Vacuum Manifold allows the processing of silica columns for nucleic acid purification by vacuum.
- Silica columns (NucleoSpin™ and NucleoSnap) can be attached to the upper luer slots of the manifold
- Binding and washing steps are carried out by vacuum instead of centrifugation
- Parallel processing of silica columns saves signficant time
- Enables fast processing of large sample volumes
Tekniske data
Tekniske data
| Produkttype | Stop-cock Valves |
| Format | NucleoSpin or NucleoSnap Columns |
| Til brug med (udstyr) | NucleoVac 24 Vacuum Manifold |
| Mængde | 24 Each |