Få mere at vide
Macherey-Nagel™ NucleoVac 96 Spacer Set
Mængde:
2 x 4 Spacers
Pakningsstørrelse:
1 sæt
Beskrivelse
The NucleoVac 96 Vacuum Manifold is designed for the rapid manual parallel purification of nucleic acids using the 8-well and 96-well NucleoSpin and NucleoFast kits. Time consuming centrifugation and decanting steps are eliminated by use of vacuum.
- NucleoVac 96 Vacuum Manifold consists of the manifold base and lid, a spacer set, and a waste container set.
- Spacer set includes:
- 2 spacers MTP/MULTI-96 PLATES
- 2 spacers ROUND-WELL BLOCKS
- 2 spacers SQUARE-WELL BLOCKS
- 2 spacers MICROTUBE RACKS
- For use of NucleoSpin™ 8‐well strips, Starter Set A (column holder and dummy strips) is required
Tekniske data
Tekniske data
| Produkttype | Spacer Set |
| Til brug med (udstyr) | NucleoVac 96 Vacuum Manifold |
| Inkluderer | 2 spacers each for: microtiter plates, round-well blocks, square-well blocks, or microtube racks |
| Mængde | 2 x 4 Spacers |