Learn More
Nucleoside-Analog-2, MedChemExpress Product Code.: 16407966
Nucleoside-Analog-2, MedChemExpress
Product Code. 16407966
100 mg, 100mg
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16407966
Brand: Medchem Express HY77652
MedChemExpress Nucleoside-Analog-2 is a 4'-Azidocytidine analogue against Hepatitis C virus (HCV) replication.
Nucleoside-Analog-2 is a 4'-Azidocytidine analogue against Hepatitis C virus (HCV) replication.
Specifications
Chemical Name or Material Nucleoside-Analog-2
CAS 876708-01-9
Color Gray
Purity Grade Notes Research
Molecular Formula C9H11N5O6
Quantity 100 mg
Solubility Information DMSO : 33.33 mg/mL (116.86 mM; Need ultrasonic) ∣H2O : 20 mg/mL (70.12 mM; Need ultrasonic)
SMILES O[C@@H]1[C@](CO)(N=[N+]=[N-])O[C@@H](N2C(NC(C=C2)=O)=O)[C@H]1O
Molecular Weight (g/mol) 285.21
Formula Weight 285.21
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Nucleoside-Analog-2, MedChemExpress