missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nucleolin Antibody (NCL/8068R), Alexa Fluor™ 594, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-24068AF594
This item is not returnable.
View return policy
Description
Nucleolin Monoclonal antibody specifically detects Nucleolin in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
Specifications
Nucleolin | |
Monoclonal | |
Alexa Fluor 594 | |
C23, FLJ45706, FLJ59041, nucleolin, Protein C23 | |
Recombinant full-length human Nucleolin protein | |
0.1 mL | |
Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, DNA Repair, DNA replication Transcription Translation and Splicing, Phospho Specific, Stem Cell Markers | |
4691 | |
Store at 4°C in the dark. | |
IgG Îş |
Western Blot, Immunohistochemistry (Paraffin) | |
NCL/8068R | |
50 mM Sodium Borate | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Nucleolin Antibody (NCL/8068R), Alexa Fluor™ 594, Novus Biologicals™ > 0.1 mL, Alexa Fluor 594
Spot an opportunity for improvement?Share a Content Correction