missing translation for 'onlineSavingsMsg'
Få mere at vide

Nuciferin, TRC

Artikelnummer. 30413856
Change view
Click to view available options
Mængde:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30413856 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30413856 Supplier Toronto Research Chemicals Supplier No. TRCN925100250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Nuciferin
CAS 475-83-2
Molekylær formel C19 H21 N O2
Mængde 250 mg
Synonym 4H-Dibenzo[de,g]quinoline','5,6,6a,7-tetrahydro-1,2-dimethoxy-6-methyl-','(6aR)-, 4H-Dibenzo[de,g]quinoline','5,6,6a,7-tetrahydro-1,2-dimethoxy-6-methyl-','(R)-, 6aβ-Aporphine','1,2-dimethoxy- (8CI), Nuciferine (6CI,7CI), (-)-Nuciferine, (6aR)-5,6,6a,7-Tetrahydro-1,2-dimethoxy-6-methyl-4H-dibenzo[de,g]quinoline, (R)-1,2-Dimethoxyaporphine, (R)-Nuciferine, 1,2-Dimethoxy-6aβ-aporphine, D-(-)-Nuciferine, Nuciferin, Sanjoinine E, VLT 049, l-Nuciferine
InChI formel InChI=1S/C19H21NO2/c1-20-9-8-13-11-16(21-2)19(22-3)18-14-7-5-4-6-12(14)10-15(20)17(13)18/h4-7,11,15H,8-10H2,1-3H3/t15-/m1/s1
SMIL COc1cc2CCN(C)[C@@H]3Cc4ccccc4c(c1OC)c23
IUPAC navn (6aR)-1,2-dimethoxy-6-methyl-5,6,6a,7-tetrahydro-4H-dibenzo[de,g]quinoline
Molekylvægt (g/mol) 295.38
Formel vægt 295.1572
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.