Få mere at vide
STEMCELL Technologies NU7441
Beskrivelse
- NU7441 is an inhibitor of DNA-dependent protein kinase (DNA-PK), an enzyme involved in the non-homologous end joining (NHEJ) DNA repair pathway
- It is highly selective for DNA-PK, with an IC50 of 14 nM (Leahy et al.)
- Reduces the frequency of NHEJ and increases the efficiency of homology-directed repair (HDR) in CRISPR-Cas9 genome editing (Robert et al.)
- Sensitizes human cancer cell lines to DNA double-strand-break-inducing therapies (chemo- or radio-therapy) by inhibiting DNA-PK activity and delaying the repair of double-strand breaks (Ciszewski et al.; Shaheen et al.; Yang et al.; Zhao et al.)
Tekniske data
Tekniske data
| CAS | 503468-95-9 |
| Synonym | KU57788 |
| Mængde | 5 mg |
| Molekylær formel | C25H19NO3S |
| Renhed | ≥98% |