Få mere at vide
Tocris Bioscience™ NTRC 824
GREENER_CHOICE
Potent and selective non-peptide NTS2 antagonist
Brand:  Tocris Bioscience™ 5438/10
Encompass
Produktkode 15736836
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter »
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
NTRC 824 is a potent and selective non-peptide neurotensin (NTS) type-2 receptor antagonist (IC50 = 38 nM). Exhibits >150-fold selectivity for NTS2 over NTS1.
Tekniske data
Tekniske data
N-[2-[5-[[(4-Methylphenyl)sulfonyl]amino]-3-(2,2,2-trifluoroacetyl)-1H-indol-1-yl]acetyl]-L-leucine
1623002-61-8
10 mg
Produktforslag
Produktforslag
Videoer
SDS
Dokumenter
Dokumenter
Certifikater
Kampagner
Kampagner