missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NTN1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103150
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total Netrin-1.
Netrin-1 is a protein target that plays a crucial role in various biological processes, particularly in the development and guidance of neuronal cells. It belongs to the netrin family of axon guidance molecules and is involved in mediating axon guidance, cell migration, and tissue morphogenesis during embryonic development. Netrin-1 functions by binding to specific receptors on the cell surface and providing directional cues for cellular movement and connectivity. Dysregulation of Netrin-1 has been implicated in various diseases, including cancer, neurological disorders, and cardiovascular conditions.
Specifications
NTN1 | |
Polyclonal | |
Unconjugated | |
NTN1 | |
AI561871; BC019633; Epididymis tissue protein Li 131P; HGNC:8029; netrin 1; netrin 1, mouse, homolog of; Netrin1; netrin-1; Ntn1; NTN1L | |
Rabbit | |
Affinity chromatography | |
RUO | |
114523, 18208, 9423 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O09118, O95631, Q924Z9 | |
NTN1 | |
A synthesized peptide derived from human NTN1(Accession O95631), corresponding to amino acid residues A572-A604. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Invitrogen™ NTN1 Polyclonal Antibody > 100 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction