Få mere at vide
Novus Biologicals™ NT5C2 Overexpression Lysate
Artikelnummer. 18273993 Shop alle Bio Techne produkter
Skift visning
Mængde:
0.1 mg
Artikelnummer. Mængde
18273993 0.1 mg
1 options
Artikelnummer. 18273993 Leverandør Novus Biologicals™ Leverandørnr. NBL113821
 Begrænset vare
Forventet forsendelse: 09-10-2026
The NT5C2 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
TRUSTED_SUSTAINABILITY
Tekniske data
Cellelinje The lysate was created in HEK293T cells, using plasmid ID RC2194 and based on accession number NM_12229. The protein contains a C-terminal DDK tag.
Til brug med (applikation) Western Blot
Mængde 0.1 mg
Opbevaringsbuffer RIPA buffer
Renhed Protein
Testspecificitet Homo sapiens 5'-nucleotidase, cytosolic II (NT5C2), transcript variant 1, mRNA.
Indhold og opbevaring -80°C. Avoid freeze-thaw cycles.
Forskningskategori Proteases & Other Enzymes
Proteintilstand Native
For Research Use Only