Få mere at vide
Nt-Methyl-d3-histamine Dihydrochloride, TRC
Artikelnummer. 30467287
Skift visning
Mængde:
0.5 mg
Pakningsstørrelse:
0.5mg
Artikelnummer. Mængde unitSize
30467287 0.5 mg 0.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30467287 Leverandør Toronto Research Chemicals Leverandørnr. TRCM3121540.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Nt-Methyl-d3-histamine Dihydrochloride
CAS 344299-49-6
Molekylær formel C6 D3 H8 N3 . 2 Cl H
Mængde 0.5 mg
Synonym Nt-Methyl-d3-histamine Dihydrochloride, 1H-Imidazole-4-ethanamine','1-(methyl-d3)-','dihydrochloride (9CI), 2-[1-(Methyl-d3)-1H-imidazol-4-yl]ethan-1-amine hydrochloride (1:2), 2-[1-(Methyl-d3)-1H-imidazol-4-yl]ethan-1-amine dihydrochloride, 1-Methylhistamine-d3 dihydrochloride, 1-Methyl-d3-histamine dihydrochloride, 1-Methylhistamine-D3 (N-methyl-D3) dihydrochloride
InChI formel InChI=1S/C6H11N3.2ClH/c1-9-4-6(2-3-7)8-5-9;;/h4-5H,2-3,7H2,1H3;2*1H/i1D3;;
SMIL Cl.Cl.[2H]C([2H])([2H])n1cnc(CCN)c1
IUPAC navn 2-[1-(trideuteriomethyl)imidazol-4-yl]ethanamine;dihydrochloride
Molekylvægt (g/mol) 201.112
Formel vægt 200.0675